α-(1-Ethylpropyl)-3-(methylthio)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210267-1G 210267-500MG 210267-5G
77466-740EA 497.49 USD
77466-740 77479-472 77489-756
α-(1-Ethylpropyl)-3-(methylthio)benzenemethanol ≥95%
α-(1-Ethylpropyl)-3-(methylthio)benzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀OS
MW: 224.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34867032
CAS Number: 2153035-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR