α-(1-Ethylpropyl)-4-methoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

210254-1G 210254-500MG 210254-5G 210254-25G
77466-736EA 297.67 USD
77466-736 77479-468 77489-752 77500-052
α-(1-Ethylpropyl)-4-methoxybenzenemethanol ≥95%
α-(1-Ethylpropyl)-4-methoxybenzenemethanol

Precaución: For research use only

Formula: C₁₃H₂₀O₂
MW: 208.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12786935
CAS Number: 860586-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR