1-Propanone, 1-(3-ethoxyphenyl)- ≥95%

Supplier: AOBChem USA

Warning

209961-1G 209961-500MG 209961-5G 209961-25G
77466-728EA 341.96 USD
77466-728 77479-460 77489-742 77500-044
1-Propanone, 1-(3-ethoxyphenyl)- ≥95%
1-Propanone, 1-(3-ethoxyphenyl)-

Precaución: For research use only

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617668
CAS Number: 1183722-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR