1-(3,5-Dimethylphenyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

209953-1G 209953-500MG 209953-5G 209953-25G
77466-724EA 248.23 USD
77466-724 77479-456 77489-738 77500-042
1-(3,5-Dimethylphenyl)-1-propanone ≥95%
1-(3,5-Dimethylphenyl)-1-propanone

Precaución: For research use only

Formula: C₁₁H₁₄O
MW: 162.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617656
CAS Number: 92821-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR