(2-(Benzylthio)phenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

20908-1G 20908-500MG 20908-5G 20908-25G
77466-712EA 370.8 USD
77466-712 77479-444 77489-722 77500-024
(2-(Benzylthio)phenyl)boronic acid ≥95%
(2-(Benzylthio)phenyl)boronic acid

Precaución: For research use only

Formula: C₁₃H₁₃BO₂S
Storage Temperature: Ambient
MDL Number: MFCD12024308
CAS Number: 1221446-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR