B,B'-(2,3,5,6-Tetrafluoro-1,4-phenylene)bis[boronic acid] ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (Perfluoro-1,4-phenylene)diboronic acid

208781-1G 208781-500MG 208781-5G 208781-25G
77466-708EA 122.57 USD
77466-708 77479-440 77489-716 77500-018
B,B'-(2,3,5,6-Tetrafluoro-1,4-phenylene)bis[boronic acid] ≥95%
B,B'-(2,3,5,6-Tetrafluoro-1,4-phenylene)bis[boronic acid]

Precaución: For research use only

Formula: C₆H₄B₂F₄O₄
MW: 237.71 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31726380
CAS Number: 1380435-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR