4,5-Dimethyl-2-fluoropyridine ≥95%

Supplier: AOBChem USA

Warning

20835-1G 20835-500MG 20835-5G 20835-25G
77466-704EA 397.58 USD
77466-704 77479-436 77489-710 77500-012
4,5-Dimethyl-2-fluoropyridine ≥95%
4,5-Dimethyl-2-fluoropyridine

Precaución: For research use only

Formula: C₇H₈FN
Storage Temperature: Ambient
MDL Number: MFCD16610911
CAS Number: 1227602-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR