1,2-Dichloro-4-(trifluoromethyl)-3-(trimethylsilyl)-benzene ≥95%

Supplier: AOBChem USA

Warning

20687-1G 20687-500MG 20687-5G 20687-25G
77466-700EA 415.09 USD
77466-700 77479-432 77489-706 77500-008
1,2-Dichloro-4-(trifluoromethyl)-3-(trimethylsilyl)-benzene ≥95%
1,2-Dichloro-4-(trifluoromethyl)-3-(trimethylsilyl)-benzene

Precaución: For research use only

Formula: C₁₀H₁₁Cl₂F₃Si
MW: 287.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30178765
CAS Number: 871254-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR