5-Fluoro-2,3-dihydro-1,4-benzodioxin-6-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

20613-1G 20613-500MG 20613-5G
77466-696EA 597.4 USD
77466-696 77479-428 77489-702
5-Fluoro-2,3-dihydro-1,4-benzodioxin-6-carboxylic acid ≥95%
5-Fluoro-2,3-dihydro-1,4-benzodioxin-6-carboxylic acid

Precaución: For research use only

Formula: C₉H₇FO₄
MW: 198.148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977035
CAS Number: 524005-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR