2-Chloro-5-bromo-6-fluorobenzamide ≥95%

Supplier: AOBChem USA

Warning

20474-1G 20474-500MG 20474-5G 20474-25G
77466-680EA 305.91 USD
77466-680 77479-412 77489-684 77499-996
2-Chloro-5-bromo-6-fluorobenzamide ≥95%
2-Chloro-5-bromo-6-fluorobenzamide

Precaución: For research use only

Formula: C₇H₄BrClFNO
Storage Temperature: Ambient
MDL Number: MFCD20484395
CAS Number: 702640-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR