1-(3-Chloro-4-fluorophenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(3-Chloro-4-fluorophenyl)-1-propanone, 3'-Chloro-4'-fluoropropiophenone

204656-1G 204656-5G 204656-25G
77466-676EA 60.77 USD
77466-676 77489-680 77499-992
1-(3-Chloro-4-fluorophenyl)propan-1-one ≥95%
1-(3-Chloro-4-fluorophenyl)propan-1-one

Precaución: For research use only

Formula: C₉H₈ClFO
MW: 186.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143013
CAS Number: 194943-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR