3-Fluoro-2-methoxybenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

20445-1G 20445-500MG 20445-5G 20445-25G
77466-672EA 72.1 USD
77466-672 77479-406 77489-676 77499-990
3-Fluoro-2-methoxybenzotrifluoride ≥97%
3-Fluoro-2-methoxybenzotrifluoride

Precaución: For research use only

Formula: C₈H₆F₄O
MW: 194.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185639
CAS Number: 1214351-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR