3-(3-Methylphenyl)-2-propynoic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-(m-Tolyl)propiolic acid

203520-1G 203520-500MG 203520-5G 203520-25G
77466-668EA 118.45 USD
77466-668 77479-402 77489-672 77499-986
3-(3-Methylphenyl)-2-propynoic acid ≥95%
3-(3-Methylphenyl)-2-propynoic acid

Precaución: For research use only

Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14707282
CAS Number: 29835-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR