3-(4-Methylphenyl)-2-propynoic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-(p-Tolyl)propiolic acid

203517-1G 203517-500MG 203517-5G 203517-25G
77466-664EA 144.2 USD
77466-664 77479-398 77489-668 77499-982
3-(4-Methylphenyl)-2-propynoic acid ≥95%
3-(4-Methylphenyl)-2-propynoic acid

Precaución: For research use only

Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11934439
CAS Number: 2227-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR