1-[2-(4-Methylphenyl)ethynyl]cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

203324-1G 203324-500MG 203324-5G
77466-660EA 576.8 USD
77466-660 77479-394 77489-664
1-[2-(4-Methylphenyl)ethynyl]cyclopentanol ≥95%
1-[2-(4-Methylphenyl)ethynyl]cyclopentanol

Precaución: For research use only

Formula: C₁₄H₁₆O
MW: 200.28 g/mol
Storage Temperature: Ambient
CAS Number: 350711-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR