1-[2-(4-Fluorophenyl)ethynyl]cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

203224-1G 203224-500MG 203224-5G
77466-656EA 546.93 USD
77466-656 77479-390 77489-660
1-[2-(4-Fluorophenyl)ethynyl]cyclopentanol ≥95%
1-[2-(4-Fluorophenyl)ethynyl]cyclopentanol

Precaución: For research use only

Formula: C₁₃H₁₃FO
MW: 204.24 g/mol
Storage Temperature: Ambient
CAS Number: 647843-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR