2-(3-Chloro-2,6-difluorophenyl)butan-2-ol ≥95%

Supplier: AOBChem USA

Warning

203033-1G 203033-500MG 203033-5G 203033-25G
77466-652EA 415.09 USD
77466-652 77479-386 77489-652 77499-972
2-(3-Chloro-2,6-difluorophenyl)butan-2-ol ≥95%
2-(3-Chloro-2,6-difluorophenyl)butan-2-ol

Precaución: For research use only

Formula: C₁₀H₁₁ClF₂O
MW: 220.644 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34865696
CAS Number: 2969389-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR