(3-Chloro-2,6-difluorophenyl)(cyclohexyl)methanol ≥95%

Supplier: AOBChem USA

Warning

202939-1G 202939-500MG 202939-5G 202939-25G
77466-648EA 415.09 USD
77466-648 77479-382 77489-648 77499-968
(3-Chloro-2,6-difluorophenyl)(cyclohexyl)methanol ≥95%
(3-Chloro-2,6-difluorophenyl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₃H₁₅ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD34865602
CAS Number: 2969351-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR