(6-Chloro-2-fluoro-3-methoxyphenyl)(cyclohexyl)methanol ≥95%

Supplier: AOBChem USA

Warning

202930-1G 202930-500MG 202930-5G 202930-25G
77466-644EA 415.09 USD
77466-644 77479-378 77489-644 77499-964
(6-Chloro-2-fluoro-3-methoxyphenyl)(cyclohexyl)methanol ≥95%
(6-Chloro-2-fluoro-3-methoxyphenyl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₈ClFO₂
MW: 272.743 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34865593
CAS Number: 2969388-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR