(6-Bromo-3-ethoxy-2-fluorophenyl)(cyclohexyl)methanol ≥97%

Supplier: AOBChem USA

Warning

202924-1G 202924-500MG 202924-5G 202924-25G
77466-640EA 415.09 USD
77466-640 77479-374 77489-640 77499-960
(6-Bromo-3-ethoxy-2-fluorophenyl)(cyclohexyl)methanol ≥97%
(6-Bromo-3-ethoxy-2-fluorophenyl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₅H₂₀BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD34865587
CAS Number: 2969369-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR