(2-Bromo-6-fluoro-4-methylphenyl)(cyclohexyl)methanol ≥95%

Supplier: AOBChem USA

Warning

202920-1G 202920-500MG 202920-5G 202920-25G
77466-636EA 415.09 USD
77466-636 77479-370 77489-636 77499-956
(2-Bromo-6-fluoro-4-methylphenyl)(cyclohexyl)methanol ≥95%
(2-Bromo-6-fluoro-4-methylphenyl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD34865583
CAS Number: 2969471-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR