(3-Bromo-2-chloro-6-fluorophenyl)(cyclohexyl)methanol ≥95%

Supplier: AOBChem USA

Warning

202916-1G 202916-500MG 202916-5G 202916-25G
77466-632EA 415.09 USD
77466-632 77479-366 77489-632 77499-952
(3-Bromo-2-chloro-6-fluorophenyl)(cyclohexyl)methanol ≥95%
(3-Bromo-2-chloro-6-fluorophenyl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₃H₁₅BrClFO
Storage Temperature: Ambient
MDL Number: MFCD34865579
CAS Number: 2969351-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR