(2-Bromo-3-chloro-6-fluorophenyl)(cyclohexyl)methanol ≥95%

Supplier: AOBChem USA

Warning

202914-1G 202914-500MG 202914-5G 202914-25G
77466-628EA 415.09 USD
77466-628 77479-362 77489-628 77499-948
(2-Bromo-3-chloro-6-fluorophenyl)(cyclohexyl)methanol ≥95%
(2-Bromo-3-chloro-6-fluorophenyl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₃H₁₅BrClFO
Storage Temperature: Ambient
MDL Number: MFCD34865577
CAS Number: 2969471-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR