(5-Bromo-3-ethoxy-2-fluorophenyl)(cyclohexyl)methanol ≥95%

Supplier: AOBChem USA

Warning

202897-1G 202897-500MG 202897-5G 202897-25G
77466-624EA 415.09 USD
77466-624 77479-358 77489-624 77499-944
(5-Bromo-3-ethoxy-2-fluorophenyl)(cyclohexyl)methanol ≥95%
(5-Bromo-3-ethoxy-2-fluorophenyl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₅H₂₀BrFO₂
MW: 331.221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34865560
CAS Number: 2969369-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR