(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)(cyclohexyl)methanol ≥95%

Supplier: AOBChem USA

Warning

202877-1G 202877-500MG 202877-5G 202877-25G
77466-620EA 415.09 USD
77466-620 77479-354 77489-620 77499-940
(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)(cyclohexyl)methanol ≥95%
(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₉H₂₀BrFO
MW: 363.264 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34865540
CAS Number: 2969471-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR