2-Fluoro-5-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

20281-1G 20281-500MG 20281-5G 20281-25G
77466-616EA 80.34 USD
77466-616 77479-350 77489-616 77499-936
2-Fluoro-5-(methylthio)benzaldehyde ≥97%
2-Fluoro-5-(methylthio)benzaldehyde

Precaución: For research use only

Formula: C₈H₇FOS
MW: 170.204 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26687655
CAS Number: 310466-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR