4-Carbamoyl-3,5-difluorobenzeneboronic acid pinacol ester ≥95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2,6-Difluoro-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)benzamide
20201-1G
20201-500MG
20201-5G
20201-25G
77466-608EA
183.34
USD
77466-608
77479-342
77489-608
77499-930
4-Carbamoyl-3,5-difluorobenzeneboronic acid pinacol ester ≥95%
4-Carbamoyl-3,5-difluorobenzeneboronic acid pinacol ester
Precaución: For research use only
Formula:
C₁₃H₁₆BF₂NO₃ MW: 283.08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17215212 CAS Number: 957345-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...