4-Fluoro-2-methyl-1-(methylsulfanyl)benzene ≥97%

Supplier: AOBChem USA

Warning

20197-1G 20197-5G 20197-25G
77466-604EA 60.77 USD
77466-604 77489-604 77499-926
4-Fluoro-2-methyl-1-(methylsulfanyl)benzene ≥97%
4-Fluoro-2-methyl-1-(methylsulfanyl)benzene

Precaución: For research use only

Formula: C₈H₉FS
MW: 156.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617797
CAS Number: 1314931-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR