1-(phenylethynyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

201823-1G 201823-500MG 201823-5G
77466-600EA 594.31 USD
77466-600 77479-336 77489-598
1-(phenylethynyl)cyclopentanol ≥95%
1-(phenylethynyl)cyclopentanol

Precaución: For research use only

Formula: C₁₃H₁₄O
MW: 186.2496 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00060797
CAS Number: 25118-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR