1-Bromo-2-(3-methoxypropoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

200410-1G 200410-5G 200410-25G
77466-588EA 64.89 USD
77466-588 77489-582 77499-906
1-Bromo-2-(3-methoxypropoxy)benzene ≥97%
1-Bromo-2-(3-methoxypropoxy)benzene

Precaución: For research use only

Formula: C₁₀H₁₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD12762613
CAS Number: 1079402-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR