6-Chloro-4-fluoro-3-pyridinecarboxylic acid ≥96%

Supplier: AOBChem USA

Warning

199704-1G 199704-500MG 199704-5G
77466-580EA 1405.95 USD
77466-580 77479-322 77489-574
6-Chloro-4-fluoro-3-pyridinecarboxylic acid ≥96%
6-Chloro-4-fluoro-3-pyridinecarboxylic acid

Precaución: For research use only

Formula: C₆H₃ClFNO₂
MW: 175.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13189250
CAS Number: 1060809-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR