1-Bromo-2,4-dimethoxy-3-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

19960-1G 19960-500MG 19960-5G 19960-25G
77466-576EA 364.62 USD
77466-576 77479-318 77489-570 77499-896
1-Bromo-2,4-dimethoxy-3-(methoxymethoxy)benzene ≥95%
1-Bromo-2,4-dimethoxy-3-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₁₀H₁₃BrO₄
Storage Temperature: Ambient
MDL Number: MFCD34755757
CAS Number: 2901085-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR