2-Iodo-3-(trifluoromethoxy)bromobenzene ≥97%

Supplier: AOBChem USA

Warning

19928-1G 19928-500MG 19928-5G 19928-25G
77466-572EA 250.29 USD
77466-572 77479-314 77489-566 77499-892
2-Iodo-3-(trifluoromethoxy)bromobenzene ≥97%
2-Iodo-3-(trifluoromethoxy)bromobenzene

Precaución: For research use only

Formula: C₇H₃BrF₃IO
MW: 366.902 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18398073
CAS Number: 1060691-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR