3,4-Difluoro-5-methylphenol ≥95%

Supplier: AOBChem USA

Warning

198687-1G 198687-500MG 198687-5G
77466-568EA 925.97 USD
77466-568 77479-310 77489-562
3,4-Difluoro-5-methylphenol ≥95%
3,4-Difluoro-5-methylphenol

Precaución: For research use only

Formula: C₇H₆F₂O
MW: 144.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28790488
CAS Number: 1806289-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR