2-Chloro-4-(trifluoromethyl)benzoic acid ethyl ester ≥95%

Supplier: AOBChem USA

Warning

19798-1G 19798-500MG 19798-5G 19798-25G
77466-564EA 129.78 USD
77466-564 77479-306 77489-556 77499-882
2-Chloro-4-(trifluoromethyl)benzoic acid ethyl ester ≥95%
2-Chloro-4-(trifluoromethyl)benzoic acid ethyl ester

Precaución: For research use only

Formula: C₁₀H₈ClF₃O₂
MW: 252.618 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12923258
CAS Number: 192805-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR