(3,5-Dimethylphenyl)phenylmethanone ≥95%

Supplier: AOBChem USA

Warning

197003-1G 197003-500MG 197003-5G
77466-556EA 499.55 USD
77466-556 77479-298 77489-546
(3,5-Dimethylphenyl)phenylmethanone ≥95%
(3,5-Dimethylphenyl)phenylmethanone

Precaución: For research use only

Formula: C₁₅H₁₄O
Storage Temperature: Ambient
CAS Number: 13319-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR