3-Bromo-2-chloro-5-isopropylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

19655-1G 19655-500MG 19655-5G 19655-25G
77466-548EA 339.9 USD
77466-548 77479-290 77489-536 77499-864
3-Bromo-2-chloro-5-isopropylbenzaldehyde ≥95%
3-Bromo-2-chloro-5-isopropylbenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₀BrClO
MW: 261.543 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902561
CAS Number: 2586126-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR