(4-((4-methoxybenzyl)oxy)-2-methylphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

196471-1G 196471-500MG 196471-5G 196471-25G
77466-544EA 415.09 USD
77466-544 77479-286 77489-532 77499-860
(4-((4-methoxybenzyl)oxy)-2-methylphenyl)boronic acid ≥95%
(4-((4-methoxybenzyl)oxy)-2-methylphenyl)boronic acid

Precaución: For research use only

Formula: C₁₅H₁₇BO₄
Storage Temperature: Ambient
MDL Number: MFCD34863830
CAS Number: 2639372-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR