2-Chloro-4-[(trifluoromethyl)thio]phenol ≥96%

Supplier: AOBChem USA

Warning

195979-1G 195979-500MG 195979-5G
77466-540EA 751.9 USD
77466-540 77479-282 77489-528
2-Chloro-4-[(trifluoromethyl)thio]phenol ≥96%
2-Chloro-4-[(trifluoromethyl)thio]phenol

Precaución: For research use only

Formula: C₇H₄ClF₃OS
MW: 228.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25966444
CAS Number: 70783-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR