4,5-Dimethylpyridine-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

19559-1G 19559-500MG 19559-5G
77466-532EA 578.86 USD
77466-532 77479-274 77489-520
4,5-Dimethylpyridine-3-carboxylic acid ≥97%
4,5-Dimethylpyridine-3-carboxylic acid

Precaución: For research use only

Formula: C₈H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD13181799
CAS Number: 878794-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR