1-Bromo-3-fluoro-4-methyl-2-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

195295-1G 195295-500MG 195295-5G
77466-528EA 696.28 USD
77466-528 77479-270 77489-516
1-Bromo-3-fluoro-4-methyl-2-(trifluoromethyl)benzene ≥95%
1-Bromo-3-fluoro-4-methyl-2-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₈H₅BrF₄
MW: 257.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34863380
CAS Number: 2169466-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR