1-Fluoro-2-iodo-3-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

194654-1G 194654-500MG 194654-5G
77466-520EA 557.23 USD
77466-520 77479-262 77489-508
1-Fluoro-2-iodo-3-(trifluoromethyl)benzene ≥95%
1-Fluoro-2-iodo-3-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₇H₃F₄I
MW: 290 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22490776
CAS Number: 110192-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR