2-(4-Bromo-2,5-dimethylphenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

19456-1G 19456-500MG 19456-5G 19456-25G
77466-516EA 363.59 USD
77466-516 77479-258 77489-504 77499-846
2-(4-Bromo-2,5-dimethylphenyl)-1,3-dioxolane ≥95%
2-(4-Bromo-2,5-dimethylphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₁H₁₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD31619403
CAS Number: 2221812-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR