2-Chloro-4-fluoro-3-(methylthio)-1-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

194091-1G 194091-500MG 194091-5G 194091-25G
77466-512EA 415.09 USD
77466-512 77479-254 77489-500 77499-842
2-Chloro-4-fluoro-3-(methylthio)-1-(trifluoromethyl)benzene ≥95%
2-Chloro-4-fluoro-3-(methylthio)-1-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₈H₅ClF₄S
MW: 244.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34862948
CAS Number: 2710205-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR