1-(4-Bromo-5-methyl-2-thienyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

193792-1G 193792-500MG 193792-5G
77466-508EA 659.2 USD
77466-508 77479-250 77489-496
1-(4-Bromo-5-methyl-2-thienyl)-2,2-difluoroethanone ≥95%
1-(4-Bromo-5-methyl-2-thienyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₇H₅BrF₂OS
MW: 255.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30742174
CAS Number: 1694575-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR