5-Bromo-1,3-difluoro-2-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

19318-1G 19318-5G 19318-25G
77466-504EA 66.95 USD
77466-504 77489-490 77499-838
5-Bromo-1,3-difluoro-2-(methoxymethoxy)benzene ≥97%
5-Bromo-1,3-difluoro-2-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₈H₇BrF₂O₂
MW: 253.041 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31619402
CAS Number: 623148-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR