5-Chloro-4-methylpyridine-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

19301-1G 19301-500MG 19301-5G 19301-25G
77466-500EA 419.21 USD
77466-500 77479-244 77489-484 77499-832
5-Chloro-4-methylpyridine-2-carboxylic acid ≥97%
5-Chloro-4-methylpyridine-2-carboxylic acid

Precaución: For research use only

Formula: C₇H₆ClNO₂
MW: 171.581 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07375101
CAS Number: 886365-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR