5-Bromo-1-chloro-2-(cyclopropylmethoxy)-3-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

192621-1G 192621-500MG 192621-5G 192621-25G
77466-496EA 415.09 USD
77466-496 77479-240 77489-480 77499-828
5-Bromo-1-chloro-2-(cyclopropylmethoxy)-3-fluorobenzene ≥95%
5-Bromo-1-chloro-2-(cyclopropylmethoxy)-3-fluorobenzene

Precaución: For research use only

Formula: C₁₀H₉BrClFO
MW: 279.533 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34862123
CAS Number: 2969432-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR