3-Bromo-4-ethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

192505-1G 192505-500MG 192505-5G 192505-25G
77466-492EA 113.3 USD
77466-492 77479-236 77489-474 77499-824
3-Bromo-4-ethylbenzoic acid ≥97%
3-Bromo-4-ethylbenzoic acid

Precaución: For research use only

Formula: C₉H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD03819559
CAS Number: 99548-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR