4-(methylsulfonyl)morpholine ≥97%

Supplier: AOBChem USA

Warning

192453-1G 192453-500MG 192453-5G 192453-25G
77466-488EA 122.57 USD
77466-488 77479-232 77489-470 77499-820
4-(methylsulfonyl)morpholine ≥97%
4-(methylsulfonyl)morpholine

Precaución: For research use only

Formula: C₅H₁₁NO₃S
MW: 165.2107 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00091634
CAS Number: 1697-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR